missing translation for 'onlineSavingsMsg'
Learn More

MP BIOMEDICALS Magbeads FastRNA Kit Ready-to-Use for MPure-32 96 Preps

Magbeads FastRNA Kit is intended for simple and rapid extraction of total RNA from tissue and culture cells samples using the MPure-32™ aNAP System.

Brand:  MP BIOMEDICALS 117033400


Product Code. 30067476

  • £1090.00 / 96 preps

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

Magbeads FastRNA Kit combines the speed and efficiency of silica-based technology with the convenient handling of magnetic particles for purification of total RNA.

  • Samples are lysed and RNA is purified from lysates in one step through its binding to the silica surface of the particles in the presence of a chaotropic salt.
  • The particles are separated from the lysates using a magnet and DNA is removed by treatment with RNase-free DNase.
  • The magnetic particles are efficiently washed, and RNA is eluted in RNase-free water.

The kit is based on superparamagnetic particles purification technology, no phenolchloroform extraction or alcohol precipitation. The whole extraction process is only 60 minutes. Purified RNA is ready for downstream applications such as RT-PCR, virus RNA testing and so on. The kits buffers can be used for both manual extraction process and automatic nucleic acid extraction machines.

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Tissue and Cells in Culture
RNA Extraction
RNA Kit
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Product Identifier
  • (MPure-32) Magbeads FastRNA Kit

Signal Word
  • Danger

Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 3
  • Respiratory sensitiser Category 1
  • Skin corrosion/irritation Category 1 B
  • Skin corrosion/irritation Category 2

Hazard Statement
  • H226-Flammable liquid and vapour.
  • H302-Harmful if swallowed.
  • H314-Causes severe skin burns and eye damage.
  • H315-Causes skin irritation.
  • H318-Causes serious eye damage.
  • H319-Causes serious eye irritation.
  • H332-Harmful if inhaled.
  • H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.

Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P264-Wash thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P321-Specific treatment (see on this label).
  • P370+P378-In case of fire: Use CO2, powder or waterspray to extinguish.
  • P405-Store locked up.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.

Supplemental information
  • MIXTURE LIST-Contains : Proteinase K, Guanidine Thiocyanate, Ethanol

SDS
Documents

Documents

Certificates
Special Offers

Special Offers