missing translation for 'onlineSavingsMsg'
Learn More

MP Biomedicals™ MagBeads FastRNA™ Kit for Virus, 50 preps

The MagBeads FastRNA™ Kit for Virus is designed to quickly and effectively isolate high-quality total viral RNA from cell culture media, swabs, and bodily fluids such as blood, plasma, serum, saliva, and more in 60 minutes.

Brand:  MP Biomedicals™ 116578050


Product Code. 30067473

  • £192.00 / 50 preps

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

  • High-quality viral RNA extraction from a variety of samples
  • Compatible with cell culture media, swabs, and bodily fluids
  • Magnetic bead-based method designed for manual and automated workflows
  • Flexible mid- to high-throughput processing
  • No phenol chloroform extractions or toxic reagents
  • Less than 60 minutes to process samples
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Store Carrier RNA and Proteinase K at 2 to 8 °C. Store other kit components at room temperature, Dry environment.
1.2 mL Magnetic Beads VRM, 32 mL Lysis Buffer VRM, 16.5 mL Wash Buffer VRM 1, 6 mL Wash Buffer VRM 2, 10 mL RNase-free Water, 1 vial Carrier RNA, 1 vial Proteinase K, 1.5 mL Proteinase K Buffer, 2 x 96-deep well plates
50 Preps
Cell Culture Media, Swabs, and Bodily Fluids
RNA Extraction Kit
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Product Identifier
  • MagBeads FastRNA Kit for Virus

Signal Word
  • Danger

Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Respiratory sensitiser Category 1
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3

Hazard Statement
  • H302-Harmful if swallowed.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
  • H335-May cause respiratory irritation.

Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P264-Wash thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P321-Specific treatment (see on this label).
  • P405-Store locked up.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.

Supplemental information
  • MIXTURE LIST-Contains : Guanidine Hydrochloride, Proteinase K

SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Unless specified otherwise, MP Biomedical's products are for research or further manufacturing use only, not for direct human use.