missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MP Biomedicals™ Magbeads FastRNA Kit
MagBeads FastRNA Kit is intended for simple and rapid extraction of total RNA from tissue and culture cells samples.
Brand: MP Biomedicals™ 116572096
Additional Details : Weight : 0.68000kg
Description
- Intended for simple and rapid extraction of total RNA from tissue and culture cells samples
- Based on superparamagnetic particles purification technology, no phenol-chloroform extraction or alcohol precipitation
- The whole extraction process is only 60 minutes. Purified RNA is ready for downstream applications such as RTPCR, virus RNA testing and so on
- The kits buffers can be used for both manual extraction process, automatic nucleic acid extraction machines
Specifications
Kit | |
Mini | |
DNA and RNA Sequencing |
Magnetic Bead | |
Tissue, Cell Cultures | |
96 Preps |