missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MAGEA6 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 14602-1-AP-20UL
This item is not returnable.
View return policy
Description
MAGEA6 is a member of the MAGEA gene family. The members of this family have their entire coding sequences located in the last exon, and the encoded proteins show 50 to 80% sequence identity between each other. The promoters and first exons of the MAGEA genes show considerable variability, suggesting that the existence of this gene family enables the same function to be expressed under different transcriptional controls. The MAGEA genes are expressed at a high level in a number of tumors of various histologic types, and are silent in normal tissues with the exception of testis and placenta. The MAGEA genes are clustered on chromosome Xq28. They may be implicated in some hereditary disorders, such as dyskeratosis congenita.Specifications
MAGEA6 | |
Polyclonal | |
Unconjugated | |
MAGEA6 | |
Cancer/testis antigen 1.6, CT1.6, MAGE 3b, MAGE 6 antigen, MAGE3B, MAGE3B antigen, MAGE6, MAGEA6, melanoma antigen family A, 6, Melanoma associated antigen 6 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
17142, 4105 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Immunofluorescence, Immunocytochemistry, Western Blot | |
0.18 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P43360 | |
Magea6 | |
MAGEA6 Fusion Protein Ag6148 | |
20 μL | |
Primary | |
Mouse, Human | |
Antibody | |
IgG |