missing translation for 'onlineSavingsMsg'
Learn More

MAGI1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Brand:  Proteintech 55048-1-AP-150UL

 View more versions of this product

Product Code. 16826485

  • £349.00 / 150µL

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

The antibody is specific to MAGI1. The antibody can recognize all isoforms of MAGI1.

The protein encoded by this gene is a member of the membrane-associated guanylate kinase homologue (MAGUK) family. MAGUK proteins participate in the assembly of multiprotein complexes on the inner surface of the plasma membrane at regions of cell-cell contact. The product of this gene may play a role as scaffolding protein at cell-cell junctions. Alternatively spliced transcript variants encoding different isoforms have been identified.
Specifications

Specifications

MAGI1
Polyclonal
Unconjugated
MAGI1
AIP 3, AIP3, BAI1 associated protein 1, BAIAP1, BAP 1, BAP1, MAGI 1, MAGI1, TNRC19, WW domain containing protein 3, WWP3
Rabbit
Antigen Affinity Chromatography
RUO
14924, 500261, 9223
-20°C
Liquid
Western Blot
0.74 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q4L1J4, Q6RHR9, Q96QZ7
Magi1
Peptide
150 μL
Primary
Human, Mouse, Rat
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers