Learn More
Invitrogen™ MagicMedia™ E. coli Expression Medium, Powder
£52.25 - £221.00
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15351532
|
Invitrogen™
K6810 |
1 L |
£52.25
1kg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15398136
|
Invitrogen™
K6815 |
5 x 1 L |
£221.00
1 set |
Estimated Shipment: 12-11-2024
Log in to see stock. |
|||||
Description
MagicMedia E. coli Expression Medium is specifically designed to dramatically increase the yield of recombinant proteins in T7-regulated E. coli expression systems without monitoring culture OD (optical density) or adding inducer. The proprietary MagicMedia formulation enables E. coli growth to reach culture densities 3- to 10-fold higher than those achieved with traditional LB + IPTG methods, allowing you to:• Achieve 3–10 times higher protein yields (Figure 1) with much less hands-on time
• Eliminate time-consuming OD monitoring steps—simply inoculate, grow the culture overnight, and harvest (Figure 2)
• Reduce culture volume to simplify handling and scale-up processes
• Ideal for simultaneous expression of multiple clones for high-throughput applications
MagicMedia E. coli Expression Medium, powder, consists of a proprietary Component A (in foil pouch powder format) and Component B (in buffer liquid format).
MagicMedia E. coli Expression Medium is also available in a ready-to-use, pre-sterilized liquid format (Cat. No. K6803).
Specifications
Escherichia coli | |
Kit | |
MagicMedia™ |
For Research Use Only. Not for use in diagnostic procedures.