Learn More
Applied Biosystems™ MagMAX™ Cell-Free Total Nucleic Acid Isolation Kit
The MagMAX Cell-Free Total Nucleic Acid Isolation Kit is designed for enrichment of all nucleic acid species present in cell-free biological samples such as plasma.
Brand: Applied Biosystems™ A36716
646.00 GBP valid until 2024-12-20
Use promo code "21651" to get your promotional price.
Description
The MagMAX Cell-Free Total Nucleic Acid Isolation Kit is designed for enrichment of all nucleic acid species present in cell-free biological samples such as plasma. The kit utilizes MagMAX magnetic-bead technology, ensuring reproducible recovery of high-quality nucleic acid that is suitable for a broad range of applications, including real-time PCR, digital PCR, and next-generation sequencing.
The magnetic bead–based purification format allows you to easily process a wide range of sample input volumes (1 mL to 6 mL). When used with the KingFisher Duo Prime or KingFisher Flex Magnetic Particle Processor, 6 to 24 plasma samples of 2-mL input volume can be isolated at once. Alternatively, samples can be processed manually with the use of a magnetic stand.
Features of the MagMAX Cell-Free Total Nucleic Acid Isolation Kit include:
- Automation-ready, phenol-free extraction
- Fast procedure allows 6 to 24 samples to be processed in 90 minutes or less when used with the KingFisher Duo Prime or KingFisher Flex Magnetic Particle Processor with a 24 Deep Well Head
- Flexible protocol accommodates sample volume inputs ranging from 1 mL to 6 mL of plasma
- No need for carrier RNA
- Elution volumes ranging from 15 μL to 60 μL
- Compatible with the Oncomine Lung cfTNA Research Assay
Go beyond cell-free DNA and get the total picture
Isolating and analyzing circulating cell-free total nucleic acid (cfTNA) from plasma and other cell-free samples serves as a non-invasive method to detect and monitor targets associated with certain disease states such as cancer.
The first step in attaining this valuable information is efficient nucleic acid isolation that specifically recovers the circulating nucleic acid species that contains information related to the disease state. The importance of cell-free DNA (cfDNA) is becoming established, but it may not tell the whole story. Cell-free RNA (cfRNA) sequences serve as critical biomarkers as well in the form of microRNAs and gene fusions. This inclusion of the cfRNA with the cfDNA provides an opportunity for researchers to expand their studies and advance human health.
The MagMAX Cell-Free Total Nucleic Acid kit is developed by the same Thermo Fisher Scientific scientists that brought you the MagMAX Cell-Free DNA Isolation Kit. The difference is the inclusion of all cfRNA species in the eluate, allowing you to increase the amount of information available from a single cell-free sample. Just like the cfDNA Isolation Kit, the cfTNA kit is designed for both manual and automated isolation of nucleic acid from a wide sample volume input range of 1 to 6 mL. The phenol-free, magnetic-bead format makes the kit fully amenable to automated isolation.
Thanks to the scalable format that allows for various sample inputs, it is possible to vary the number of samples processed with a single kit. For example, enough reagents are provided to process 50 samples of 2 mL plasma each. Alternatively, with the same kit it is possible to process 25 samples of 4 mL plasma each. Higher and lower input volumes are possible and the number of samples that can be processed will vary accordingly. This unique product attribute allows you to tailor the product to your specific needs.
Benefits of MagMAX magnetic bead–based purification
Magnetic beads offer many benefits compared to other technologies for isolating nucleic acid. Beads bind the DNA more efficiently than glass-fiber filters, resulting in higher and more consistent yields. Additionally, because filters and vacuum manifolds are not used, there is no risk of filter clogging due to cellular particulates in samples. This clogging issue is of particular concern with protein-rich, large-volume samples such as plasma that are commonly used in circulating cell-free nucleic acid studies.
Specifications
• 175 mL MagMAX Cell-Free Total Nucleic Acid lysis/binding solution • 100 mL (final volume) MagMAX Cell-Free Total Nucleic Acid wash solution concentrate • 21 mL MagMAX Cell-Free Total Nucleic Acid elution solution • 3.5 mL MagMAX Cell-Free Total Nucleic Acid magnetic beads • 1.5 mL MagMAX Cell-Free Total Nucleic Acid proteinase K |
|
Magnetic Bead | |
Next-Generation Sequencing, Real-Time PCR | |
MagMAX™ | |
1 kit | |
90 min. |
Kit | |
Plasma | |
High-throughput Compatible, Manual Protocols, Automated Protocols | |
Total Nucleic Acid Isolation Kit | |
Room Temperature |
For Research Use Only. Not for use in diagnostic procedures.