missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Applied Biosystems™ MagMAX™ Cell and Tissue DNA Extraction Buffer
MagMAX DNA Cell and Tissue Extraction Buffer, a component of the MagMAX DNA Multi-Sample Ultra 2.0 Kit with Cell and Tissue Extraction Buffer, is offered separately here for those kit users who need additional buffer.
£93.00 - £293.00
Specifications
Sample Type | Cells, Tissue |
---|---|
For Use With (Application) | Next-Generation Sequencing, PCR, Real-Time PCR |
High-throughput Compatibility | Automated Protocols, High-throughput Compatible, Manual Protocols |
Product Line | MagMAX™ |
Product Type | Cell and Tissue DNA Extraction Buffer |
Product Code | Brand | No. of Reactions | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | No. of Reactions | Price | Quantity & Availability | |||||
16584810
|
Applied Biosystems™
A45469 |
100 Preps |
£93.00
Each |
Estimated Shipment: 09-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
17165772
|
Applied Biosystems™
A45470 |
1000 Preps |
£293.00
1000 preps |
In Stock Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
MagMAX DNA Cell and Tissue Extraction Buffer, a component of the MagMAX DNA Multi-Sample Ultra 2.0 Kit with Cell and Tissue Extraction Buffer, is offered separately here for those kit users who need additional buffer.
Specifications
Cells, Tissue | |
Automated Protocols, High-throughput Compatible, Manual Protocols | |
Cell and Tissue DNA Extraction Buffer | |
Genomic DNA |
Next-Generation Sequencing, PCR, Real-Time PCR | |
MagMAX™ | |
Room Temperature |
For Research Use Only. Not for use in diagnostic procedures.