missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Applied Biosystems™ MagMAX™ Express-96 Standard Magnetic Head
Shop All Applied Biosystems Products
Click to view available options
Quantity:
Each
Unit Size:
Each
Description
Applied Biosystems™ Replacement head for the MagMAX™ Express-96 Standard Magnetic Particle Processors (Cat No. 4400076 and 4400078). One head is included.
Specifications
Specifications
| Format | 96-well Plate |
| Capacity | 96 Samples |
| Content And Storage | Store at room temperature. |
| For Use With (Equipment) | MagMAX™ Express-96 Standard Magnetic Particle Processor |
| Product Line | MagMAX |
| Quantity | Each |
| Shipping Condition | Room Temperature |
For research use only. Not for use in diagnostic procedures.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction