missing translation for 'onlineSavingsMsg'
Learn More

Applied Biosystems™ MagMAX™ FFPE DNA/RNA Ultra Kit

Product Code. 15654529
Click to view available options
Quantity:
1 kit
Unit Size:
1 set
This item is not returnable. View return policy

Product Code. 15654529

Brand: Applied Biosystems™ A31881

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

The MagMAX™ FFPE DNA/RNA Ultra Kit recovers both the DNA and RNA (including microRNA) from the same FFPE sample enabling scientists to analyze all the important biomarkers contained in the sample such as hotspot mutations, CNVs, gene fusions, and indels.

Scalable, sequential DNA & RNA isolation from FFPE. The MagMAX™ FFPE DNA/RNA Ultra Kit recovers both the DNA and RNA (including microRNA) from the same FFPE sample enabling scientists to analyze all the important biomarkers contained in the sample such as hotspot mutations, CNVs, gene fusions, and indels.

  • More information from less sample—sequentially extract DNA and RNA from the same FFPE section to advance your cancer research
  • Automation-ready technology—process 12 to 96 samples when used with Thermo Scientific™ KingFisher™ Duo Prime or KingFisher Flex Purification System
  • A flexible protocol—the kit can also isolate just RNA or just DNA from your FFPE sample and accommodates sample input ranges from 5 μM to 40 μM
TRUSTED_SUSTAINABILITY

Specifications

Content And Storage • 1 x 385 μL Protease, Store at -5 to -30°C
• 1 x 10 mL Protease Digestion Buffer, Store at Room Temperature
• 1 x 38.5 mL Binding Solution, Store at Room Temperature
• 1 x 1.95 mL Nucleic Acid Binding Beads, Store at 2°C to 8°C
• 1 x 38.5 mL DNA Wash Buffer, Store at Room Temperature
• 1 x 210 mL Wash Solution 2 Concentrate, Store at Room Temperature
• 1 x 5 mL Elution Solution, Store at Room Temperature
• 1 x 115 mL RNA Wash Buffer Concentrate, Store at Room Temperature
• 1 x 1.92 mL DNase, Store at -5 to -30°C
• 1 x 960 μL DNase Buffer, Store at -5 to -30°C
Format Kit
Isolation Technology Magnetic Bead
Sample Type FFPE, Fixed Samples
For Use With (Application) Next-Generation Sequencing, RT-PCR (Endpoint), Real-Time PCR, PCR, microRNA Analysis
High-throughput Compatibility High Throughput Compatibility
Product Line MagMAX
Product Type FFPE DNA/RNA Ultra Kit
Quantity 1 kit
Shipping Condition Approved for shipment at Room Temperature or on Wet or Dry Ice
Target Total RNA, miRNA, Total Nucleic Acids, Genomic DNA, mRNA, Transcriptome RNA
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • MagMAX FFPE DNA/RNA Ultra Kit
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 3
  • Serious eye damage/eye irritation Category 1
  • Serious eye damage/eye irritation Category 2
  • Respiratory sensitiser Category 1
  • Skin corrosion/irritation Category 1 C
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H302-Harmful if swallowed.
  • H314-Causes severe skin burns and eye damage.
  • H315-Causes skin irritation.
  • H318-Causes serious eye damage.
  • H319-Causes serious eye irritation.
  • H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
  • H412-Harmful to aquatic life with long lasting effects.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P264-Wash hands thoroughly after handling.
  • P270-Do not eat, drink or smoke when using this product.
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P284-¢In case of inadequate ventilation! wear respiratory protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P330-Rinse mouth.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
  • P342+P311-If experiencing respiratory symptoms: Call a POISON CENTER/doctor/ .
  • P362+P364-Take off contaminated clothing and wash it before reuse.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contains : Thiocyanic acid, Proteinase, serine

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.