Learn More
Applied Biosystems™ MagMAX™ Microbiome Bead Tubes
These bead tubes are included in the MagMAX Microbiome Ultra Nucleic Acid Isolation Kit.
Brand: Applied Biosystems™ A42351
102.00 GBP valid until 2024-11-30
Use promo code "21651" to get your promotional price.
This item is not returnable.
View return policy
Description
These bead tubes are included in the MagMAX Microbiome Ultra Nucleic Acid Isolation Kit (Cat. No. A42358). They are made available separately for applications that require more tubes than are provided in the kit.
Specifications
Store at room temperature. |
|
High-throughput Compatible, Manual Protocols, Automated Protocols | |
MagMAX™ | |
Room Temperature |
Bacteria, Blood, Buccal Samples, Cells, Fungi, Liquid Samples (e.g. Serum), Viral Samples, Yeast | |
Next-Generation Sequencing, RT-PCR (Endpoint), Real-Time PCR | |
100 tubes | |
Viral DNA/RNA, Pathogen DNA/RNA, Total Nucleic Acids |
For Research Use Only. Not for use in diagnostic procedures.