Learn More
Applied Biosystems™ MagMAX™ Pathogen RNA/DNA Kit
The Applied Biosystems™ MagMAX™ Pathogen RNA/DNA Kit enables purification of RNA and DNA from viruses and easy-to-lyse bacteria and parasites, using magnetic particle technology in a 96-well format.
Brand: Applied Biosystems™ 4462359
Description
PCR inhibitors are effectively removed in the process, making the resulting nucleic acid ideal for real-time PCR and RT-PCR applications.
Key Features:
- Convenience and flexibility: one kit for RNA and DNA, suitable for a wide range of sample types sample input volumes
- Increased confidence in your results: fewer false-negatives due to effective PCR inhibitor removal
- Improved workflow efficiency: process 96 samples in approximately 45 minutes using the MagMAX™ Express-96 Deep Well Magnetic Particle Processor
The MagMAX™ Pathogen RNA/DNA Kit has been optimized to provide a convenient, flexible solution. This single kit can accomodate:
- Purification of RNA and DNA
- Purification from viruses and easy-to-lyse bacteria and parasites
- Low and high sample input volumes
- A wide range of sample types:
- Low cell content samples (e.g., serum and plasma; nasal, tracheal, and cloacal swabs; ear notches)
- Whole blood
- Semen
- Oral fluid
- Feces
- Biomed Diagnostics InPouch™ TF (Tritrichomonas foetus) culture
The MagMAX™ Pathogen RNA/DNA Kit is Easy to Automate
Purify RNA/DNA from 96 samples in approximately 45 minutes using the MagMAX™ Express-96 Deep Well Magnetic Particle Processor. Low and high sample input volumes can be used for most sample types, and multiple sample types can be processed at the same time on the same plate after sample-specific pre-processing, saving time and plastics.
Increase the Accuracy of Your Results
Reduce false-negative results due to the effective PCR inhibitor removal achieved by the MagMAX™ Pathogen RNA/DNA Kit. Magnetic particles bind RNA/DNA more efficiently and are more thoroughly washed than glass fiber filters, resulting in higher yields and purity of RNA/DNA. In addition, no sample cross contamination has been observed due to extensive optimization of the MagMAX™ Express-96 Deep Well Magnetic Particle Processor programs.
Specifications
Store at room temperature: 1 bottle of Lysis⁄Binding Solution Concentrate 1 bottle of Wash Solution 1 Concentrate 2 bottles of Wash Solution 2 Concentrate 1 bottle of Elution Buffer Store at 4 °C: 1 bottle of Nucleic Acid Binding Beads Store at -20 °C: 1 bottle of Lysis ENHANCER 5 tubes of Carrier Nucleic Acid |
|
Magnetic Bead | |
Any Swab Type, Bacteria, BioMed Diagnostics InPouch™ TF Culture, Cell-free Biological Fluids, Ear Notches, Feces, Mosquito, Parasite Samples, Plasma, Saliva, Serum, Tissue Homogenates, Viral Samples, Viral Transport Media Samples, Whole Blood | |
High-throughput Compatible, Not High-throughput Compatible (Manual), Automated Protocols | |
Real-Time Quantitative PCR (qPCR), Reverse Transcriptase PCR (RT-PCR), PCR | |
MagMAX™ | |
Pathogen DNA/RNA, Viral DNA/RNA |
96-well Plate | |
45 min. | |
Viral DNA⁄RNA, Pathogen DNA⁄RNA | |
KingFisher™ Flex Magnetic Particle Processor, MagMAX™ Express-96 Deep Well Magnetic Particle Processor, MagMAX™ Express-96 Standard Magnetic Particle Processor, KingFisher 96, MagMAX™ Express Magnetic Particle Processor | |
480 preps | |
480 preps |
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.