Learn More
Applied Biosytstems™ MagMAX™ Prime Bead Beating Tubes
MagMAX Prime Bead Beating Tubes are specifically designed to enable efficient, high-throughput nucleic acid extraction from stool.
Brand: Applied Biosytstems™ A58155
Additional Details : Weight : 0.27400kg
Description
MagMAX Prime Bead Beating Tubes are specifically designed to enable efficient, high-throughput nucleic acid extraction from stool. They are designed to be used with MagMAX Prime Stool Lysis Buffer and MagMAX Prime Viral/Pathogen NA Isolation kits to maximize RNA and DNA yield.
Specifications
Magnetic | |
Store at 15–25°C. | |
Beads | |
Next-Generation Sequencing, RT-PCR (Endpoint), Real-Time PCR | |
Isolate and purify viral, bacterial, parasitic, and fungal nucleic acid | |
DNA/RNA | |
Reagent | |
400 μL | |
Viral, Bacterial, Parasitic, and Fungal Nucleic Acid |
Clear | |
Tube | |
Fecal Swab in Transport Media, Stool | |
High-throughput Compatible, Automated Protocols | |
100 Reactions | |
MagMAX™ | |
100 tubes | |
Room Temperature |