missing translation for 'onlineSavingsMsg'
Learn More

Applied Biosytstems™ MagMAX™ Prime Stool Lysis Buffer

MagMAX Prime Stool Lysis Buffer is specifically designed to enable efficient, high-throughput nucleic acid extraction from stool samples.

Brand:  Applied Biosytstems™ A58154

51.00 GBP valid until 2024-12-20
Use promo code "21651" to get your promotional price.


Product Code. 17924821

  • £60.00 / 80mL

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

MagMAX Prime Stool Lysis Buffer is specifically designed to enable efficient, high-throughput nucleic acid extraction from stool samples. It is designed to be used in conjunction with the MagMAX Prime bead-beating tubes and MagMAX Prime Viral/Pathogen NA Isolation kits to maximize RNA and DNA yield.

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Magnetic
Store at 15–25°C.
Beads
Next-Generation Sequencing, RT-PCR (Endpoint), Real-Time PCR
Isolate and purify viral, bacterial, parasitic, and fungal nucleic acid
DNA/RNA
Reagent
400 μL
Viral, Bacterial, Parasitic, and Fungal Nucleic Acid
Clear
Bottle
Fecal Swab in Transport Media, Stool
High-throughput Compatible, Automated Protocols
Up to 100 Reactions
MagMAX™
80 mL
Room Temperature
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

For Research Use Only.