Learn More
Applied Biosytstems™ MagMAX™ Prime Viral/Pathogen Binding Solution
MagMAX Prime Viral/Pathogen Binding Solution is offered separately here for applications that require more Binding Solution than is provided in the MagMAX Prime Viral/Pathogen NA Isolation kits.
Brand: Applied Biosytstems™ A58150
402.02 GBP valid until 2024-09-28
Use promo code "21626" to get your promotional price.
Alert:
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21626” to get your promotional price
Description
MagMAX Prime Viral/Pathogen Binding Solution is offered separately here for applications that require more Binding Solution than is provided in the MagMAX Prime Viral/Pathogen NA Isolation kits.
Specifications
Magnetic | |
Store at 15–25°C. | |
Beads | |
Next-Generation Sequencing, RT-PCR (Endpoint), Real-Time PCR | |
Isolate and purify viral, bacterial, parasitic, and fungal nucleic acid | |
DNA/RNA | |
Reagent | |
200–400 μL |
Clear | |
Bottle | |
Swabs in Transport Media, Saliva, Urine, Serum, Plasma | |
High-throughput Compatible, Automated Protocols | |
Up to 2000 Reactions | |
MagMAX™ | |
530 mL | |
Viral, Bacterial, Parasitic, and Fungal Nucleic Acid |
For Research Use Only.