Learn More
Applied Biosytstems™ MagMAX™ Prime Viral/Pathogen Proteinase K Dye
MagMAX Prime Viral/Pathogen Proteinase K Dye is offered separately here for applications that require more Proteinase K Dye than is provided in the MagMAX Prime Viral/Pathogen NA Isolation kits.
Brand: Applied Biosytstems™ A58152
Additional Details : Weight : 0.05000kg
Description
MagMAX Prime Viral/Pathogen Proteinase K Dye is offered separately here for applications that require more Proteinase K Dye than is provided in the MagMAX Prime Viral/Pathogen NA Isolation kits.
Specifications
Magnetic | |
Store at 15–25°C. | |
Beads | |
Next-Generation Sequencing, RT-PCR (Endpoint), Real-Time PCR | |
Isolate and purify viral, bacterial, parasitic, and fungal nucleic acid | |
DNA/RNA | |
Reagent | |
Viral, Bacterial, Parasitic, and Fungal Nucleic Acid |
Clear | |
Tube | |
Swabs in Transport Media, Saliva, Urine, Serum, Plasma | |
High-throughput Compatible, Automated Protocols | |
Up to 2000 Reactions | |
MagMAX™ | |
200–400 μL |