missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Applied Biosytstems™ MagMAX™ Prime Viral/Pathogen Wash I Solution
MagMAX Prime Viral/Pathogen Wash I Solution is offered separately here for applications that require more Wash I Solution than is provided in the MagMAX Prime Viral/Pathogen NA Isolation kits.
Brand: Applied Biosytstems™ A58149
520.20 GBP valid until 2024-12-20
Use promo code "21651" to get your promotional price.
This item is not returnable.
View return policy
Description
MagMAX Prime Viral/Pathogen Wash I Solution is offered separately here for applications that require more Wash I Solution than is provided in the MagMAX Prime Viral/Pathogen NA Isolation kits.
Specifications
Magnetic | |
Store at 15–25°C. | |
Beads | |
Next-Generation Sequencing, RT-PCR (Endpoint), Real-Time PCR | |
Isolate and purify viral, bacterial, parasitic, and fungal nucleic acid | |
DNA/RNA | |
Reagent | |
Room Temperature |
Clear | |
Bottle | |
Swabs in Transport Media, Saliva, Urine, Serum, Plasma | |
High-throughput Compatible, Automated Protocols | |
Up to 2000 Reactions | |
MagMAX™ | |
200–400 μL | |
Viral, Bacterial, Parasitic, and Fungal Nucleic Acid |
For Research Use Only.