Learn More
Applied Biosystems™ MagMAX™ Total RNA Elution Buffer
This elution buffer is included in the MagMAX-96 Total RNA Isolation Kit and MagMAX-96 Viral RNA Isolation Kit.
Brand: Applied Biosystems™ A41043
Additional Details : Weight : 0.02300kg
Description
This elution buffer is included in the MagMAX-96 Total RNA Isolation Kit (Cat. No. AM1830) and MagMAX-96 Viral RNA Isolation Kit (AM1836). It is made available here for separate purchase.
Specifications
Store at room temperature. |
|
MagMAX™ | |
Room Temperature |
For Research Use Only. Not for use in diagnostic procedures.