Learn More
Applied Biosystems™ MagMAX™ Viral/Pathogen Elution Buffer
This elution buffer is included in the MagMAX Viral/Pathogen Nucleic Acid Isolation kits and MagMAX Microbiome Ultra Nucleic Acid Isolation Kit.
Brand: Applied Biosystems™ A42364
275.34 GBP valid until 2024-06-29
Use promo code "21620" to get your promotional price.
Additional Details : Weight : 0.11000kg
Description
This elution buffer is included in the MagMAX Viral/Pathogen Nucleic Acid Isolation kits and MagMAX Microbiome Ultra Nucleic Acid Isolation Kit (Cat. No. A42352, A42356, A42357, and A42358). It is made available separately for applications that require more elution buffer than is provided in the kit.Specifications
Serum, Viral Samples, Buccal Samples, Blood, Cells, Bacteria, Fungi, Yeast | |
Store at room temperature. |
|
High-throughput Compatible, Manual Protocols, Automated Protocols | |
Viral DNA⁄RNA, Pathogen DNA⁄RNA, Total Nucleic Acids |
100 mL | |
Next-Generation Sequencing, RT-PCR (Endpoint), Real-Time PCR | |
MagMAX™ | |
Room Temperature |
For Research Use Only. Not for use in diagnostic procedures.