missing translation for 'onlineSavingsMsg'
Learn More

Applied Biosystems™ MagMAX™ Viral/Pathogen II (MVP II) Nucleic Acid Isolation Kit

The MagMAX Viral/Pathogen II (MVP II) Nucleic Acid Isolation Kit recovers RNA and DNA from virus and gram-negative bacteria in samples such as blood, swabs, urine, and transport media.

Brand:  Applied Biosystems™ A48383R

2126.25 GBP valid until 2024-11-30
Use promo code "21651" to get your promotional price.



Product Code. 17103791

This item is not returnable. View return policy

Description

Description

  • Automation-ready protocols designed for the KingFisher Flex and KingFisher Duo Prime systems
  • Fast procedure allows for 96 samples to be processed in <30 minutes
  • Flexible protocol accommodates sample volume inputs from 200 μL to 2 mL of biofluids or transfer media
  • No need for carrier RNA
  • Elution volumes ranging from 50 to 100 μL
  • The optimized reagents included allow you to maximize the amount of sample input
  • Up to 400 μL of sample can be processed per well in a KingFisher 96-deep well plate.
  • Protocols are available to process up to 2 mL of sample per well in a KingFisher 24-deep well plate
TRUSTED_SUSTAINABILITY
Specifications

Specifications

• 550 mL Binding Solution, store at room temperature
• 1000 mL Wash Solution, store at room temperature
• 100 mL Elution Solution, store at room temperature
• 10 mL Proteinase K, store at room temperature
• 20 mL DNA/RNA Binding Beads, store at room temperature

30 min.
MagMAX™
Room Temperature
96 Deep-well Plate, 24 Deep-well Plate
2000 Preps
2000 preps
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Product Identifier
  • MagMAX Viral/Pathogen II (MVP II) Nucleic Acid Isolation Kit

Signal Word
  • Danger

Hazard Category
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 3
  • Serious eye damage/eye irritation Category 2
  • Respiratory sensitiser Category 1
  • Skin corrosion/irritation Category 1 C
  • Skin corrosion/irritation Category 2

Hazard Statement
  • H302-Harmful if swallowed.
  • H314-Causes severe skin burns and eye damage.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
  • H412-Harmful to aquatic life with long lasting effects.

Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P264-Wash hands thoroughly after handling.
  • P270-Do not eat, drink or smoke when using this product.
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P284-¢In case of inadequate ventilation! wear respiratory protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P330-Rinse mouth.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P342+P311-If experiencing respiratory symptoms: Call a POISON CENTER/doctor/ .
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.

Supplemental information
  • MIXTURE LIST-Contains : Thiocyanic acid, acrylamide, proteinase

SDS
Documents

Documents

Certificates
Special Offers

Special Offers

For Research Use Only. Not for use in diagnostic procedures.