Learn More
Applied Biosystems™ MagMAX™ Viral/Pathogen Nucleic Acid Isolation Kit
The MagMAX Viral/Pathogen Nucleic Acid Isolation Kit is designed to recover RNA and DNA from virus and gram-negative bacteria in samples such as blood, swabs, urine, and viral transport media (VTM).
Brand: Applied Biosystems™ A42352
418.00 GBP valid until 2024-12-31
Use promo code "21969" to get your promotional price.
Additional Details : Weight : 0.39000kg Transport : UN number : 1760 Chem class : 8 Pack group : III
Description
The MagMAX Viral/Pathogen Nucleic Acid Isolation Kit is designed to recover RNA and DNA from virus and gram-negative bacteria in samples such as blood, swabs, urine, and viral transport media (VTM). The kit utilizes MagMAX magnetic-bead technology, ensuring reproducible recovery of high-quality nucleic acid compatible with a broad range of applications, including real-time PCR, digital PCR, and next-generation sequencing.
Features of the MagMAX Viral/Pathogen Nucleic Acid Isolation Kit include:
• Automation-ready protocols designed for the KingFisher Flex and KingFisher Duo Prime systems
• Fast procedure allows for 96 samples to be processed in under 30 minutes
• Flexible protocol accommodates sample volume inputs from 200 μL to 2 mL of biofluids or transfer media
• No need for carrier RNA
• Elution volumes ranging from 50 to 100 μL
Maximum sample input in a 96-deep well or 24-deep well plate format
The optimized reagents included in the MagMAX Viral/Pathogen kit allow you to maximize the amount of sample input, thereby increasing the amount of RNA and/or DNA recovered. Up to 400 μL of sample can be processed per well in a KingFisher 96-deep well plate. For larger sample volumes, protocols are available to process up to 2 mL of sample per well in a KingFisher 24-deep well plate.
Benefits of MagMAX magnetic bead–based purification
Magnetic beads offer many benefits compared to other technologies for isolating nucleic acid. Beads bind the nucleic acid more efficiently than glass-fiber filters, resulting in higher and more consistent yields. Additionally, because filters and vacuum manifolds are not used, there is no risk of filter clogging due to cellular particulates in samples. This clogging issue is of particular concern with protein-rich, large-volume samples such as whole blood or plasma that are commonly used for viral testing.
The Dynabeads MyOne Silane contained in the MagMAX Viral/Pathogen Total Nucleic Acid Isolation Kit have an optimized silica-like surface chemistry with a highly specific surface area that allows efficient kinetics and high sensitivity in nucleic acid capture. These aspects make Dynabeads MyOne Silane ideal for the recovery viral RNA and DNA present at low concentrations in body fluids. Once captured, these nucleic acids can then be eluted in 50 μL of elution buffer for use in downstream applications.
Specifications
• 55 mL Binding Solution, store at room temperature • 100 mL Wash Solution, store at room temperature • 10 mL Elution Solution, store at room temperature • 1 mL Proteinase K, store at room temperature • 2 mL DNA/RNA Binding Beads, store at room temperature |
|
<30 min. | |
Up to 200 Preps |
96 Deep-well Plate, 24 Deep-well Plate | |
MagMAX™ | |
Room Temperature |
Safety and Handling
- MagMAX Viral/Pathogen Nucleic Acid Isolation Kit
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 4
- LONG-TERM AQUATIC HAZARD Chronic 3
- Serious eye damage/eye irritation Category 1
- Serious eye damage/eye irritation Category 2
- Respiratory sensitiser Category 1
- Skin corrosion/irritation Category 1 C
- Skin corrosion/irritation Category 2
Hazard Statement
- H302-Harmful if swallowed.
- H314-Causes severe skin burns and eye damage.
- H315-Causes skin irritation.
- H318-Causes serious eye damage.
- H319-Causes serious eye irritation.
- H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
- H412-Harmful to aquatic life with long lasting effects.
Precautionary Statement
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P264-Wash hands thoroughly after handling.
- P270-Do not eat, drink or smoke when using this product.
- P273-Avoid release to the environment.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P284-¢In case of inadequate ventilation! wear respiratory protection.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P302+P352-IF ON SKIN: Wash with plenty of water/soap.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
- P330-Rinse mouth.
- P332+P313-If skin irritation occurs: Get medical advice/attention.
- P342+P311-If experiencing respiratory symptoms: Call a POISON CENTER/doctor/ .
- P363-Wash contaminated clothing before reuse.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
- MIXTURE LIST-Contains : Thiocyanic acid, Acrylamide, Proteinase
For Research Use Only. Not for use in diagnostic procedures.