Learn More
Magnesium 1,1,1-trifluoro-2,4-pentanedionate dihydrate, 98%, Thermo Scientific™
£39.68 - £158.00
Chemical Identifiers
CAS | 240131-46-8 |
---|---|
Molecular Formula | C10H12F6MgO6 |
Molecular Weight (g/mol) | 366.50 |
MDL Number | MFCD00077561,MFCD00150843 |
InChI Key | OWJDZQKGGDMWCY-UHFFFAOYSA-L |
Synonym | magnesium trifluoroacetylacetonate dihydrate, magnesium1,1,1-trifluoro-2,4-pentanedionate, magnesium 1,1,1-trifluoro-2,4-pentanedionate 2-hydrate, 3z-1,1,1-trifluoro-4-2z-5,5,5-trifluoro-4-oxopent-2-en-2-yl oxy magnesio oxy pent-3-en-2-one dihydrate |
PubChem CID | 91872989 |
IUPAC Name | magnesium;(Z)-5,5,5-trifluoro-4-oxopent-2-en-2-olate;dihydrate |
SMILES | O.O.[Mg++].CC([O-])=CC(=O)C(F)(F)F.CC([O-])=CC(=O)C(F)(F)F |
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
240131-46-8 | |
366.50 | |
OWJDZQKGGDMWCY-UHFFFAOYSA-L | |
91872989 | |
O.O.[Mg++].CC([O-])=CC(=O)C(F)(F)F.CC([O-])=CC(=O)C(F)(F)F |
C10H12F6MgO6 | |
MFCD00077561,MFCD00150843 | |
magnesium trifluoroacetylacetonate dihydrate, magnesium1,1,1-trifluoro-2,4-pentanedionate, magnesium 1,1,1-trifluoro-2,4-pentanedionate 2-hydrate, 3z-1,1,1-trifluoro-4-2z-5,5,5-trifluoro-4-oxopent-2-en-2-yl oxy magnesio oxy pent-3-en-2-one dihydrate | |
magnesium;(Z)-5,5,5-trifluoro-4-oxopent-2-en-2-olate;dihydrate |
Specifications
240131-46-8 | |
MFCD00077561,MFCD00150843 | |
OWJDZQKGGDMWCY-UHFFFAOYSA-L | |
magnesium;(Z)-5,5,5-trifluoro-4-oxopent-2-en-2-olate;dihydrate | |
91872989 | |
98% | |
Magnesium 1,1,1-trifluoro-2,4-pentanedionate dihydrate |
C10H12F6MgO6 | |
magnesium trifluoroacetylacetonate dihydrate, magnesium1,1,1-trifluoro-2,4-pentanedionate, magnesium 1,1,1-trifluoro-2,4-pentanedionate 2-hydrate, 3z-1,1,1-trifluoro-4-2z-5,5,5-trifluoro-4-oxopent-2-en-2-yl oxy magnesio oxy pent-3-en-2-one dihydrate | |
O.O.[Mg++].CC([O-])=CC(=O)C(F)(F)F.CC([O-])=CC(=O)C(F)(F)F | |
366.50 | |
366.50 (330.47 Anhydrous) | |
Powder |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Warning