missing translation for 'onlineSavingsMsg'
Learn More

Magnesium, 99.9+%, (trace metal basis), turnings

Product Code. 10368090 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
500 g
2.5 kg
10 kg
Unit Size:
10kg
2.5kg
500g
This item is not returnable. View return policy

Product Code. 10368090

Brand: Thermo Scientific Acros undefined

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 7439-95-4 | Mg | 24.30 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

For ultra trace analysis
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7439-95-4
Molecular Formula Mg
Molecular Weight (g/mol) 24.30
MDL Number MFCD00085308
InChI Key JLVVSXFLKOJNIY-UHFFFAOYSA-N
Synonym magnesio, sheet, powdered, magnesio italian, turnings, rieke's active, ribbon, unii-i38zp9992a, hsdb 654, nitrate, acs
PubChem CID 5462224
ChEBI CHEBI:25107
IUPAC Name magnesium
SMILES [Mg]

Specifications

Melting Point 651.0°C
Boiling Point 1107.0°C
Density 1.74g/mL
Quantity 500 g
Flash Point 500°C
Packaging Plastic bottle
Assay Percent Range 99.9+%
Form Turnings
Linear Formula Mg
Fieser 01,627; 02,254; 03,189; 04,315; 05,419; 06,351; 07,218; 10,251; 12,290; 13,170; 15,120; 16,198
Formula Weight 24.3
Percent Purity ≥99.9%
Grade Trace Metal Basis
Chemical Name or Material Magnesium, 99.9+%
Show More Show Less
compliance-icons
Product Identifier
  • Magnesium
Signal Word
  • Danger
Hazard Category
  • Flammable solid Category 1
  • Substance which\, in contact with water\, emits flammable gases Category 2
  • Self-heating substance Category 2
Hazard Statement
  • H228-Flammable solid.
  • H252-Self-heating in large quantities; may catch fire.
  • H261-In contact with water releases flammable gases.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P223-Do not allow contact with water.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.