missing translation for 'onlineSavingsMsg'
Learn More
Magnesium, 99.9+%, (trace metal basis), turnings, Thermo Scientific Chemicals Product Code.: 10574131

Magnesium, 99.9+%, (trace metal basis), turnings, Thermo Scientific Chemicals

Product Code. 10574131
2.5 kg
Click to view available options
Quantity:
500 g
2.5 kg
10 kg
This item is not returnable. View return policy

Product Code. 10574131

Brand: Thermo Scientific Acros 191080025

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

For ultra trace analysis
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7439-95-4
Molecular Formula Mg
Molecular Weight (g/mol) 24.30
MDL Number MFCD00085308
InChI Key JLVVSXFLKOJNIY-UHFFFAOYSA-N
Synonym magnesio, sheet, powdered, magnesio italian, turnings, rieke's active, ribbon, unii-i38zp9992a, hsdb 654, nitrate, acs
PubChem CID 5462224
ChEBI CHEBI:25107
IUPAC Name magnesium
SMILES [Mg]

Specifications

Melting Point 651.0°C
Boiling Point 1107.0°C
Density 1.74g/mL
Assay Percent Range 99.9+%
Linear Formula Mg
Quantity 2.5 kg
Fieser 01,627; 02,254; 03,189; 04,315; 05,419; 06,351; 07,218; 10,251; 12,290; 13,170; 15,120; 16,198
Flash Point 500°C
Formula Weight 24.3
Percent Purity ≥99.9%
Grade Trace Metal Basis
Packaging Plastic drum
Form Turnings
Chemical Name or Material Magnesium, 99.9+%
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0466322 Certificate TypeCertificate of Analysis Date20/09/2024 Catalog Number191080100,191080025,191085000
Lot NumberA0452858 Certificate TypeCertificate of Analysis Date02/05/2024 Catalog Number191080100,191080025,191085000
Lot NumberA0461406 Certificate TypeCertificate of Analysis Date22/03/2024 Catalog Number191080100,191080025,191085000
Lot NumberA0445241 Certificate TypeCertificate of Analysis Date25/08/2023 Catalog Number191080100,191080025,191085000
Lot NumberA0404148 Certificate TypeCertificate of Analysis Date13/12/2022 Catalog Number191080100,191080025,191085000

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Magnesium, 99.9+%, (trace metal basis), turnings, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.