missing translation for 'onlineSavingsMsg'
Learn More
Magnesium, AAS standard solution, Specpure™ Mg 1000μg/mL, Thermo Scientific Chemicals Product Code.: 11336359

Magnesium, AAS standard solution, Specpure™ Mg 1000μg/mL, Thermo Scientific Chemicals

Product Code. 11336359
500 mL
Click to view available options
Quantity:
100 mL
500 mL
This item is not returnable. View return policy

Product Code. 11336359

Brand: Thermo Scientific Alfa Aesar 088077.AP

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7439-95-4, 7697-37-2, 7732-18-5
Molecular Formula Mg
Molecular Weight (g/mol) 24.30
MDL Number MFCD00085308
InChI Key JLVVSXFLKOJNIY-UHFFFAOYSA-N
PubChem CID 5462224
ChEBI CHEBI:25107
IUPAC Name magnesium
SMILES [Mg]

Specifications

Quantity 500 mL
Vapor Pressure 17mm Hg at 20°C
Physical Form Liquid
Color Colorless
Concentration or Composition (by Analyte or Components) Nitric acid: 5%; Magnesium: 0.1%; Water: 94.9%
Chemical Name or Material Magnesium

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number1525580 Certificate TypeCertificate of Analysis Date17/03/2025 Catalog Number088077.AP,088077.AE
Lot Number1537417 Certificate TypeCertificate of Analysis Date11/03/2025 Catalog Number088077.AP,088077.AE
Lot Number1523208 Certificate TypeCertificate of Analysis Date11/03/2025 Catalog Number088077.AP,088077.AE
Lot Number1493495 Certificate TypeCertificate of Analysis Date15/10/2024 Catalog Number088077.AP,088077.AE
Lot Number1493602 Certificate TypeCertificate of Analysis Date15/10/2024 Catalog Number088077.AP,088077.AE

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Magnesium, AAS standard solution, Specpure™ Mg 1000μg/mL, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.