missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Magnesium Aluminum Zinc wire, 3.18mm (0.125 in.) dia., 90cm (35 in.) long, Thermo Scientific™
£91.82 - £432.00
Specifications
Chemical Name or Material | Magnesium Aluminum Zinc wire |
---|---|
Name Note | 3.18mm (0.125 in.) dia., 90cm (35 in.) long |
Avg. Mol. Wt. or Mol. Wt. Range | Mg:Al:Zn; 93:6:1 wt% |
MDL Number | MFCD01074901 |
Solubility Information | Insoluble in water. |
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
Magnesium Aluminum Zinc wire | |
Mg:Al:Zn; 93:6:1 wt% | |
Insoluble in water. | |
≈12.44g/90cm |
3.18mm (0.125 in.) dia., 90cm (35 in.) long | |
MFCD01074901 | |
Odorless | |
Wire |
Safety and Handling
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures