missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Magnesium carbonate, for biochemistry, specified according to the requirements of US, Thermo Scientific Chemicals
Click to view available options
Quantity:
250 g
1 kg
Packaging:
Plastic bottle
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For biochemistry
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
Color | White |
Physical Form | Powder |
Quantity | 250 g |
Linear Formula | MgCO3 |
Solubility Information | Solubility in water: slightly soluble |
Formula Weight | 84.31 |
Percent Purity | 40.0 to 43.5% (MgO) |
Grade | Biochemical |
Identification | conform |
Packaging | Plastic bottle |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'