Learn More
Magnesium chloride, 1M aq. soln., sterile-filtered, Thermo Scientific Chemicals
Description
Magnesium chloride, 1M aq. soln., sterile is used as an excellent magnesium ion source for synthesis and biology research. Magnesium chloride is widely used to supply the magnesium ion in various molecular biology applications, including PCR reactions and buffers. It was used to prepare buffer to lyse cells for immunoprecipitation1 and to prepare serum-free medium for electrophysiological studies of rat brain slice cultures.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Magnesium chloride, 1M aq. soln., sterile is used as an excellent magnesium ion source for synthesis and biology research. Magnesium chloride is widely used to supply the magnesium ion in various molecular biology applications, including PCR reactions and buffers. It was used to prepare buffer to lyse cells for immunoprecipitation1 and to prepare serum-free medium for electrophysiological studies of rat brain slice cultures.
Solubility
Soluble in water (542 mg/ml at 20°C), alcohol, methanol (15.5 g/100g at 0°C), and ethanol (3.61 g/100g at 0°C).
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Very hygroscopic.
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
Physical Form | Liquid |
Quantity | 250 mL |
Linear Formula | MgCl2 |
Solubility Information | Soluble in water (542mg/ml at 20°C),alcohol,methanol (15.5g/100g at 0°C),and ethanol (3.61g/100g at 0°C). |
IUPAC Name | magnesium(2+) dichloride |
Formula Weight | 95.22 |
Concentration or Composition (by Analyte or Components) | 1M aq. soln. |
Chemical Name or Material | Magnesium chloride, sterile |
RUO – Research Use Only