missing translation for 'onlineSavingsMsg'
Learn More

Magnesium Chloride Hexahydrate, ≥99.0%, Reagent Grade, Honeywell™

Product Code. 15697910
Click to view available options
Quantity:
500 g
1 kg
5 kg
Packaging:
Poly bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

Reagent Grade,≥99.0%

  • ≥95% for general laboratory use.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7791-18-6
Molecular Formula Cl2H12MgO6
Molecular Weight (g/mol) 203.30
MDL Number MFCD00149781
InChI Key DHRRIBDTHFBPNG-UHFFFAOYSA-L
IUPAC Name magnesium(2+) hexahydrate dichloride
SMILES O.O.O.O.O.O.[Mg++].[Cl-].[Cl-]

Specifications

Melting Point 117°C
Color Colorless
pH 5.0 to 6.5 (at 20°C)
Physical Form Crystalline
Quantity 1 kg
Linear Formula MgCl2 · 6H2O
UN Number NONH for all modes of transport
Formula Weight 203.3g/mol
Percent Purity ≥99.0%
Odor Odorless
Packaging Poly bottle
Density 1.57g/cu.cm at 20°C
Chemical Name or Material Magnesium chloride hexahydrate
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'