missing translation for 'onlineSavingsMsg'
Learn More

Magnesium chloride hexahydrate, ACS, 99.0-102.0%, Thermo Scientific Chemicals

Product Code. 11323278
Click to view available options
Quantity:
500 g
2 kg

Utilized in the polymerase chain reactions (PCR). Used for dust control, ice control, and road stabilization. Employed in a variety of applications such as fertilizer, mineral supplement for animals, waste water treatment, artificial seawater, textiles, paper, fireproofing agents, cements, and fire extinguishers. Used in several medicines and topical or skin related applications. Acts as a coagulant in the preparation of tofu from soy milk. Acts as an anesthetic for cephalopods.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7791-18-6
Molecular Formula Cl2H12MgO6
Molecular Weight (g/mol) 203.30
MDL Number MFCD00149781
InChI Key DHRRIBDTHFBPNG-UHFFFAOYSA-L
IUPAC Name magnesium(2+) hexahydrate dichloride
SMILES O.O.O.O.O.O.[Mg++].[Cl-].[Cl-]

Specifications

Melting Point 117°C (decomposition)
Physical Form Crystalline
Quantity 2 kg
Assay Percent Range 99.0 to 102.0%
Merck Index 14,5662
Solubility Information Highly soluble in water and alcohol. Slightly soluble in acetone and pyridine.
Formula Weight 203.30 (95.22 Anhydrous)
Percent Purity 99.0 to 102.0%
Grade ACS
Odor Odorless
Sensitivity Hygroscopic
Density 1.570 g/mL
Chemical Name or Material Magnesium chloride hexahydrate, For analysis ACS
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'