Learn More
Magnesium chloride hexahydrate, ACS, 99.0-102.0%, Thermo Scientific Chemicals
£46.50 - £86.50
Chemical Identifiers
CAS | 7791-18-6 |
---|---|
Molecular Formula | Cl2H12MgO6 |
Molecular Weight (g/mol) | 203.30 |
MDL Number | MFCD00149781 |
InChI Key | DHRRIBDTHFBPNG-UHFFFAOYSA-L |
IUPAC Name | magnesium(2+) hexahydrate dichloride |
SMILES | O.O.O.O.O.O.[Mg++].[Cl-].[Cl-] |
Produktcode | Marke | Quantity | Preis | Menge & Verfügbarkeit | |||||
---|---|---|---|---|---|---|---|---|---|
Produktcode | Marke | Quantity | Preis | Menge & Verfügbarkeit | |||||
11333278
|
Thermo Scientific Alfa Aesar
036226.36 |
500 g |
£46.50
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11323278
|
Thermo Scientific Alfa Aesar
036226.A3 |
2 kg |
£86.50
2kg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Beschreibung
Utilized in the polymerase chain reactions (PCR). Used for dust control, ice control, and road stabilization. Employed in a variety of applications such as fertilizer, mineral supplement for animals, waste water treatment, artificial seawater, textiles, paper, fireproofing agents, cements, and fire extinguishers. Used in several medicines and topical or skin related applications. Acts as a coagulant in the preparation of tofu from soy milk. Acts as an anesthetic for cephalopods.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemische Identifikatoren
7791-18-6 | |
203.30 | |
DHRRIBDTHFBPNG-UHFFFAOYSA-L | |
O.O.O.O.O.O.[Mg++].[Cl-].[Cl-] |
Cl2H12MgO6 | |
MFCD00149781 | |
magnesium(2+) hexahydrate dichloride |
Spezifikation
117°C (decomposition) | |
White | |
99.0 to 102.0% | |
MFCD00149781 | |
Highly soluble in water and alcohol. Slightly soluble in acetone and pyridine. | |
O.O.O.O.O.O.[Mg++].[Cl-].[Cl-] | |
203.30 | |
99.0 to 102.0% | |
Odorless | |
1.570 g/mL |
7791-18-6 | |
Crystals | |
Cl2H12MgO6 | |
14,5662 | |
DHRRIBDTHFBPNG-UHFFFAOYSA-L | |
magnesium(2+) hexahydrate dichloride | |
203.30 (95.22 Anhydrous) | |
ACS | |
Hygroscopic | |
Magnesium chloride hexahydrate, For analysis ACS |
Sicherheit und Handhabung
missing translation for 'einecsNumber' : 232-094-6
missing translation for 'rtecsNumber' : OM2975000
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only