missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Magnesium chloride hexahydrate, BAKER ANALYZED™ ACS, J.T. Baker™
43.74 GBP valid until 2024-12-31
Use promo code "23268" to get your promotional price.
Use promo code "23268" to get your promotional price.
Click to view available options
Quantity:
250 g
1 kg
Specifications
Boiling Point | 1412°C (1013 hPa) |
Melting Point | 117°C |
Quantity | 250 g |
Assay Percent Range | 99.0 to 102.0% |
Density | 1.57 g/cm³ (25°C) |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'