missing translation for 'onlineSavingsMsg'
Learn More

Magnesium Chloride Hexahydrate, Honeywell Fluka™
BIOPHARMA

Product Code. 15611270
Click to view available options
Quantity:
1 kg
2.5 kg
6 kg
6 x 1 kg
6 x 2.5 kg

Puriss., meets analytical specification of BP, FCC, European Pharmacopoeia, E511, ≤0.0001% Al, 99-101%,

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7791-18-6
Molecular Formula Cl2H12MgO6
Molecular Weight (g/mol) 203.30
MDL Number MFCD00149781
InChI Key DHRRIBDTHFBPNG-UHFFFAOYSA-L
IUPAC Name magnesium(2+) hexahydrate dichloride
SMILES O.O.O.O.O.O.[Mg++].[Cl-].[Cl-]

Specifications

Quantity 6 x 2.5 kg
Linear Formula MgCl2 · 6H2O
UN Number NONH for all modes of transport
Formula Weight 203.3
Percent Purity 99 to 101%
Grade puriss.
Chemical Name or Material Magnesium Chloride Hexahydrate, Meets analytical specification of Ph. Eur.BP, FCC
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.