missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Magnesium Chloride Hexahydrate, Honeywell Fluka™
![BIOPHARMA](/content/dam/fishersci/glyphs/sure_trace.png)
112.69 GBP valid until 2024-09-17
Use promo code "21690" to get your promotional price.
Use promo code "21690" to get your promotional price.
Click to view available options
Quantity:
1 kg
2.5 kg
6 kg
6 x 1 kg
6 x 2.5 kg
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
Quantity | 2.5 kg |
Linear Formula | MgCl2 · 6H2O |
UN Number | NONH for all modes of transport |
Formula Weight | 203.3g/mol |
Percent Purity | 99 to 101% |
Grade | puriss. |
Chemical Name or Material | Magnesium Chloride Hexahydrate, Meets analytical specification of Ph. Eur.BP, FCC |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'