Learn More
Thermo Scientific™ MgCl2 (magnesium chloride) (25 mM)
Optimize your PCR reaction conditions with this Magnesium Chloride (MgCl2) solution.
Brand: Thermo Scientific™ AB0359
21.17 GBP valid until 2024-06-28
Use promo code "21649" to get your promotional price.
Code : LE
Additional Details : Weight : 1.00000kg
Description
PCR reagents meet high quality standards to generate accurate results from even the most challenging molecular biology applications. Magnesium ions (Mg2+) are an important component of PCR reactions where they interact with the DNA template, dNTPs, and Taq DNA Polymerase.
Thermo Scientific PCR reagents meet high quality standards to generate accurate results from even the most challenging molecular biology applications. Magnesium ions (Mg2+) are an important component of PCR reactions where they interact with the DNA template, dNTPs, and Taq DNA Polymerase.Specifications
Magnesium Chloride | |
Liquid | |
25 mM |
Contains: • 5 mL 25 mM MgCl2 Store at -20°C. |
|
Taq DNA Polymerase | |
5 mL |
For Research Use Only. Not for use in diagnostic procedures.