missing translation for 'onlineSavingsMsg'
Learn More

Magnesium chloride, pure, Thermo Scientific Chemicals

Product Code. 13498478
Click to view available options
Quantity:
100 g
1 kg
2.5 kg
25 kg
Packaging:
Fibre drum
Plastic bottle

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7786-30-3
Molecular Formula Cl2Mg
Molecular Weight (g/mol) 95.21
MDL Number MFCD00011106
InChI Key TWRXJAOTZQYOKJ-UHFFFAOYSA-L
Synonym magnesium chloride, magnesium chloride anhydrous, mgcl2, magnesium chloride, anhydrous, magnesium 2+ ion dichloride, chloromagnesite, magnesiumchlorid, magnesium ii chloride
PubChem CID 5360315
ChEBI CHEBI:6636
SMILES [Mg++].[Cl-].[Cl-]

Specifications

Boiling Point 1412.0°C
Melting Point 708.0°C
Color White
Physical Form Powder
Quantity 25 kg
Linear Formula MgCl2
Fieser 11,31
Merck Index 15,5726
Solubility Information Solubility in water: 540g/L (20°C)
IUPAC Name magnesium(2+) dichloride
Formula Weight 95.21
Percent Purity ≥98.0% (on dry substance)
Grade Pure
Loss on Drying 1.5% max. (105°C, 2 Hrs)
Packaging Fibre drum
Chemical Name or Material Magnesium Chloride
Show More Show Less
Product Identifier
  • Magnesium chloride, anhydrous

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'