missing translation for 'onlineSavingsMsg'
Learn More

Magnesium chromate hydrate, 99.8% (metals basis), Thermo Scientific Chemicals

Product Code. 11338488
Click to view available options
Quantity:
100 g
500 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Corrosion inhibitor, pigment

Used as Corrosion inhibitor, pigment. As an ingredient in cosmetics.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 16569-85-0
Molecular Formula CrMgO4
Molecular Weight (g/mol) 140.30
MDL Number MFCD00801009
InChI Key CRGGPIWCSGOBDN-UHFFFAOYSA-N
Synonym Magnesium chromate
IUPAC Name magnesium(2+) dioxochromiumbis(olate)
SMILES [Mg++].[O-][Cr]([O-])(=O)=O

Specifications

Physical Form Crystalline
Quantity 500 g
UN Number UN2923
Solubility Information Soluble in water
Formula Weight 140.30 (Anhydrous)
Percent Purity 99.8%
Assay (metals basis)
Chemical Name or Material Magnesium chromate hydrate

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'