Learn More
Magnesium ethoxide, 98%, Thermo Scientific Chemicals
£24.30 - £184.00
Chemical Identifiers
CAS | 2414-98-4 |
---|---|
Molecular Formula | C4H10MgO2 |
Molecular Weight (g/mol) | 114.43 |
MDL Number | MFCD00009060 |
InChI Key | XDKQUSKHRIUJEO-UHFFFAOYSA-N |
Synonym | magnesium ethoxide, magnesium ethanolate, magnesium ethylate, magnesium diethanolate, unii-o81sa5a7l6, magnesium diethoxide, magnesium 2+ bis ethoxide, ethanol, magnesium salt, magnesiumethoxide, magnesium ii ethoxide |
PubChem CID | 164963 |
IUPAC Name | magnesium;ethanolate |
SMILES | CC[O-].CC[O-].[Mg+2] |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10204784
|
Thermo Scientific Acros
397470050 |
5 g | Glass bottle |
£24.30
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10041564
|
Thermo Scientific Acros
397471000 |
100 g | Glass bottle |
£44.62
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10674565
|
Thermo Scientific Acros
397475000 |
500 g | Glass bottle |
£184.00
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Beschreibung
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemische Identifikatoren
2414-98-4 | |
114.43 | |
XDKQUSKHRIUJEO-UHFFFAOYSA-N | |
164963 | |
CC[O-].CC[O-].[Mg+2] |
C4H10MgO2 | |
MFCD00009060 | |
magnesium ethoxide, magnesium ethanolate, magnesium ethylate, magnesium diethanolate, unii-o81sa5a7l6, magnesium diethoxide, magnesium 2+ bis ethoxide, ethanol, magnesium salt, magnesiumethoxide, magnesium ii ethoxide | |
magnesium;ethanolate |
Spezifikation
2414-98-4 | |
White to Gray | |
98% | |
(CH3CH2O)2Mg | |
magnesium ethoxide, magnesium ethanolate, magnesium ethylate, magnesium diethanolate, unii-o81sa5a7l6, magnesium diethoxide, magnesium 2+ bis ethoxide, ethanol, magnesium salt, magnesiumethoxide, magnesium ii ethoxide | |
XDKQUSKHRIUJEO-UHFFFAOYSA-N | |
magnesium;ethanolate | |
164963 | |
98% | |
Magnesium ethoxide |
270.0°C | |
Authentic | |
C4H10MgO2 | |
MFCD00009060 | |
Solubility in water: reacts | |
CC[O-].CC[O-].[Mg+2] | |
114.43 | |
114.43 | |
Powder |
Sicherheit und Handhabung
GHS H Statement
Flammable solid.
Self-heating; may catch fire.
Causes serious eye irritation.
Reacts violently with water.
GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Wear eye protection/face protection.
Keep cool.
Protect from sunlight.
Wash face,hands and any exposed skin thoroughly after handling.
If
GHS Signal Word: Danger
missing translation for 'einecsNumber' : 219-323-5
- Magnesium ethoxide
Signalwort
- Danger
Gefahrenkategorie
- Serious eye damage/eye irritation Category 2
- Flammable solid Category 1
- Self-heating substance Category 1
Gefahrenhinweis
- H319-Causes serious eye irritation.
- H251-Self-heating: may catch fire.
- H228-Flammable solid.
- EUH014-Reacts violently with water.
Sicherheitshinweis
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P235+P410-Keep cool. Protect from sunlight.
- P264-Wash thoroughly after handling.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P337+P313-If eye irritation persists: Get medical advice/attention.
RUO – Research Use Only