missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Magnesium iodide, ultra dry, 99.996% (metals basis), Thermo Scientific Chemicals
Click to view available options
Quantity:
1 g
10 g
Description
Magnesium iodide is used as reagents and intermediates in pharmaceuticals. It is useful in Baylis-Hillman reactions.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
Melting Point | 637°C (decomposition) |
Color | White |
Physical Form | Beads |
Quantity | 10 g |
Assay Percent Range | 99.996% (Metals basis) |
Solubility Information | Soluble in water(1480g/L at 18 C) |
Formula Weight | 278.12 |
Odor | Odorless |
Sensitivity | Hygroscopic; Light sensitive |
Chemical Name or Material | Magnesium iodide |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'