missing translation for 'onlineSavingsMsg'
Learn More

Magnesium iodide, ultra dry, 99.996% (metals basis), Thermo Scientific Chemicals

Catalog Number 11478400
Click to view available options
Quantity:
25 g
100 g
This item is not returnable. View return policy
10377-58-9
I2Mg
278.11
MFCD00049485
BLQJIBCZHWBKSL-UHFFFAOYSA-L
magnesium(2+) diiodide
[Mg++].[I-].[I-]
This item is not returnable. View return policy
10377-58-9
I2Mg
278.11
MFCD00049485
BLQJIBCZHWBKSL-UHFFFAOYSA-L
magnesium(2+) diiodide
[Mg++].[I-].[I-]

Light Sensitive and Hygroscopic

Magnesium iodide is used to prepare compounds for organic synthesis. It serves as a useful reagent in the Baylis-Hillman reaction. It reacts with cyclic sulfates of vic-diols in order to prepare the corresponding olefins.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 637°C (decomposition)
Color White
Physical Form Powder
Quantity 100 g
Assay Percent Range 99.996% (Metal basis)
Solubility Information Soluble in ether,alcohol and ammonia.
Formula Weight 278.12
Odor Odorless
Sensitivity Hygroscopic; Light sensitive
Appearance White powder
Chemical Name or Material Magnesium iodide, ultra dry
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'