missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Magnesium nitrate hexahydrate, for biochemistry

Product Code. 12153761
Click to view available options
Quantity:
500 g
2.5 kg
Packaging:
Plastic bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13446-18-9
Molecular Formula H12MgN2O12
Molecular Weight (g/mol) 256.40
MDL Number MFCD00149779
InChI Key MFUVDXOKPBAHMC-UHFFFAOYSA-N
Synonym magnesium nitrate hexahydrate, magnesium dinitrate hexahydrate, unii-v85k20ljmk, dusicnan horecnaty czech, v85k20ljmk, nitric acid, magnesium salt, hexahydrate, magnesium ii nitrate 1:2 , hexahydrate, magnesium 2+ ion hexahydrate dinitrate, magnesium, reference standard solution, dusicnan horecnaty
PubChem CID 202877
IUPAC Name magnesium;dinitrate;hexahydrate
SMILES O.O.O.O.O.O.[Mg++].[O-][N+]([O-])=O.[O-][N+]([O-])=O

Specifications

Boiling Point 330.0°C
Melting Point 89.0°C
Quantity 2.5 kg
Linear Formula Mg(NO3)2·6H2O
Flash Point 93.3°C
Merck Index 15,5738
Solubility Information Solubility in water: 420g/L (20°C). Other solubilities: freely soluble in alcohol, liquid nh3
Formula Weight 256.4
Grade Biochemical
Packaging Plastic bottle
Chemical Name or Material Magnesium nitrate hexahydrate
Show More Show Less
Product Identifier
  • Magnesium nitrate hexahydrate

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'