missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific Chemicals Magnesium nitrate hexahydrate, for biochemistry
£41.10 - £131.66
Chemical Identifiers
CAS | 13446-18-9 |
---|---|
Molecular Formula | H12MgN2O12 |
Molecular Weight (g/mol) | 256.40 |
MDL Number | MFCD00149779 |
InChI Key | MFUVDXOKPBAHMC-UHFFFAOYSA-N |
Synonym | magnesium nitrate hexahydrate, magnesium dinitrate hexahydrate, unii-v85k20ljmk, dusicnan horecnaty czech, v85k20ljmk, nitric acid, magnesium salt, hexahydrate, magnesium ii nitrate 1:2 , hexahydrate, magnesium 2+ ion hexahydrate dinitrate, magnesium, reference standard solution, dusicnan horecnaty |
PubChem CID | 202877 |
IUPAC Name | magnesium;dinitrate;hexahydrate |
SMILES | O.O.O.O.O.O.[Mg++].[O-][N+]([O-])=O.[O-][N+]([O-])=O |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
12163761
|
Thermo Scientific Chemicals
446835000 |
500 g | Plastic bottle |
£41.10
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
12153761
|
Thermo Scientific Chemicals
446830025 |
2.5 kg | Plastic bottle |
£131.66
2.50kg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
13446-18-9 | |
256.40 | |
MFUVDXOKPBAHMC-UHFFFAOYSA-N | |
202877 | |
O.O.O.O.O.O.[Mg++].[O-][N+]([O-])=O.[O-][N+]([O-])=O |
H12MgN2O12 | |
MFCD00149779 | |
magnesium nitrate hexahydrate, magnesium dinitrate hexahydrate, unii-v85k20ljmk, dusicnan horecnaty czech, v85k20ljmk, nitric acid, magnesium salt, hexahydrate, magnesium ii nitrate 1:2 , hexahydrate, magnesium 2+ ion hexahydrate dinitrate, magnesium, reference standard solution, dusicnan horecnaty | |
magnesium;dinitrate;hexahydrate |
Specifications
330.0°C | |
13446-18-9 | |
Mg(NO3)2·6H2O | |
93.3°C | |
magnesium nitrate hexahydrate, magnesium dinitrate hexahydrate, unii-v85k20ljmk, dusicnan horecnaty czech, v85k20ljmk, nitric acid, magnesium salt, hexahydrate, magnesium ii nitrate 1:2 , hexahydrate, magnesium 2+ ion hexahydrate dinitrate, magnesium, reference standard solution, dusicnan horecnaty | |
MFUVDXOKPBAHMC-UHFFFAOYSA-N | |
magnesium;dinitrate;hexahydrate | |
202877 | |
Biochemical |
89.0°C | |
H12MgN2O12 | |
MFCD00149779 | |
15,5738 | |
Solubility in water: 420g/L (20°C). Other solubilities: freely soluble in alcohol, liquid nh3 | |
O.O.O.O.O.O.[Mg++].[O-][N+]([O-])=O.[O-][N+]([O-])=O | |
256.40 | |
256.4 | |
Magnesium nitrate hexahydrate |
Safety and Handling
Warning
Product Identifier
- Magnesium nitrate hexahydrate
RUO – Research Use Only