missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Magnesium Nitrate Hexahydrate, Honeywell Fluka™
Click to view available options
Quantity:
100 g
500 g
1 kg
2.5 kg
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
Quantity | 500 g |
Linear Formula | Mg(NO3)2 · 6H2O |
UN Number | NONH for all modes of transport |
SMILES | O.O.O.O.O.O.[Mg++].[O-][N+]([O-])=O.[O-][N+]([O-])=O |
Formula Weight | 256.41g/mol |
Percent Purity | 99% |
Grade | ACS Reagent |
Density | Vapor Desnity: 6 (vs air) |
Chemical Name or Material | Magnesium Nitrate Hexahydrate |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'