missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Magnesium nitrate hydrate, Puratronic™, 99.999% (metals basis), Thermo Scientific Chemicals
Click to view available options
Quantity:
5 g
25 g
100 g
500 g
Description
It is used in printing, chemical, agriculture and ceramics industries. Its fertilizer grade has 10.5% nitrogen and 9.4% magnesium.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
Boiling Point | 330°C (decomposition) |
Melting Point | 89°C |
Physical Form | Crystalline Aggregates |
Quantity | 5 g |
UN Number | UN1474 |
Merck Index | 14,5674 |
Solubility Information | Freely soluble in water. Moderately soluble in ethanol and ammonia. |
Formula Weight | 148.32 (Anhydrous) |
Percent Purity | 99.999% |
Grade | Puratronic™ |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'