Learn More
Magnesium oxide, 99.95% (metals basis), Thermo Scientific Chemicals
For Vacuum Deposition
£95.10 - £1022.00
Chemical Identifiers
CAS | 1309-48-4 |
---|---|
Molecular Formula | MgO |
Molecular Weight (g/mol) | 40.30 |
MDL Number | MFCD00011109 |
InChI Key | CPLXHLVBOLITMK-UHFFFAOYSA-N |
Synonym | magnesium oxide, magnesia, seawater magnesia, causmag, granmag, maglite, periclase, seasorb, animag |
PubChem CID | 14792 |
IUPAC Name | oxomagnesium |
SMILES | O=[Mg] |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11386988
|
Thermo Scientific Alfa Aesar
014684.14 |
25 g |
£95.10
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11396988
|
Thermo Scientific Alfa Aesar
014684.22 |
100 g |
£275.00
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11376988
|
Thermo Scientific Alfa Aesar
014684.36 |
500 g |
£1022.00
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Magnesium oxide is used as a refractive material, which is physically and chemically stable at higher temperature. Magnesium oxide is a raw material for producing portland cement. It is also used in agricultural, chemical construction, environment al and other industrial applications. It is also used in medicine, colorimetry, industrial cables, heating elements, ceramics, spin-tunneling devices, and several other materials.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
1309-48-4 | |
40.30 | |
CPLXHLVBOLITMK-UHFFFAOYSA-N | |
14792 | |
O=[Mg] |
MgO | |
MFCD00011109 | |
magnesium oxide, magnesia, seawater magnesia, causmag, granmag, maglite, periclase, seasorb, animag | |
oxomagnesium |
Specifications
3600°C | |
1309-48-4 | |
Powder | |
MgO | |
magnesium oxide, magnesia, seawater magnesia, causmag, granmag, maglite, periclase, seasorb, animag | |
CPLXHLVBOLITMK-UHFFFAOYSA-N | |
oxomagnesium | |
14792 | |
1.736 | |
Magnesium oxide |
2852°C | |
White | |
99.95% (Metals basis) | |
MFCD00011109 | |
Soluble in acid and ammonia. Insoluble in alcohol. | |
O=[Mg] | |
40.30 | |
40.3 | |
3.58 g/cm3 at 20°C |
Safety and Handling
missing translation for 'einecsNumber' : 215-171-9
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only