missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Magnesium Oxide, Powder, U.S.P. - F.C.C., J.T. Baker™
Click to view available options
Quantity:
500 g
2 kg
Packaging:
Poly Bottle
Poly Pail
Specifications
Specifications
Physical Form | Powder |
Quantity | 500 g |
Formula Weight | 40.3 |
Grade | FCC/USP |
Packaging | Poly Bottle |
Specific Gravity | 1L = 3.58kg |
Chemical Name or Material | Magnesium Oxide |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'