Learn More
Magnesium oxide substrate, 10x10x1mm, polished one side, 100 orientation, Thermo Scientific Chemicals
£80.30 - £358.00
Chemical Identifiers
CAS | 1309-48-4 |
---|---|
Molecular Formula | MgO |
Molecular Weight (g/mol) | 40.30 |
MDL Number | MFCD00011109 |
InChI Key | CPLXHLVBOLITMK-UHFFFAOYSA-N |
Synonym | magnesium oxide, magnesia, seawater magnesia, causmag, granmag, maglite, periclase, seasorb, animag |
PubChem CID | 14792 |
IUPAC Name | oxomagnesium |
SMILES | O=[Mg] |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11378847
|
Thermo Scientific Alfa Aesar
038503.KT |
1 Ea. |
£80.30
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11388847
|
Thermo Scientific Alfa Aesar
038503.KU |
5 Ea. |
£358.00
Pack of 5 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Magnesium oxide substrate is used widely in high temperature superconductor. It is also used as deposition of ferro electric thin film coatings on magnesium oxide substrates. It is also growing applications for MgO in the field of plasma display panel (PDP) technology.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
1309-48-4 | |
40.30 | |
CPLXHLVBOLITMK-UHFFFAOYSA-N | |
14792 | |
O=[Mg] |
MgO | |
MFCD00011109 | |
magnesium oxide, magnesia, seawater magnesia, causmag, granmag, maglite, periclase, seasorb, animag | |
oxomagnesium |
Specifications
1309-48-4 | |
Plate | |
MFCD00011109 | |
Soluble in acid,ammonia. Insoluble in alcohol. | |
O=[Mg] | |
40.30 | |
Odorless |
White | |
MgO | |
magnesium oxide, magnesia, seawater magnesia, causmag, granmag, maglite, periclase, seasorb, animag | |
CPLXHLVBOLITMK-UHFFFAOYSA-N | |
oxomagnesium | |
14792 | |
Magnesium Oxide Substrate, Polished One Side |
Safety and Handling
missing translation for 'einecsNumber' : 215-171-9
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only