missing translation for 'onlineSavingsMsg'
Learn More

Magnesium phosphate, tribasic hydrate, 98+%, pure, Thermo Scientific Chemicals

£24.22 - £60.90

Chemical Identifiers

CAS 53408-95-0
Molecular Formula Mg3O8P2
Molecular Weight (g/mol) 262.86
MDL Number MFCD00150251
InChI Key GVALZJMUIHGIMD-UHFFFAOYSA-H
Synonym magnesium phosphate hydrate, trimagnesium diphosphate hydrate, acmc-20akl7, 3mg.2po4.h2o, trimagnesium 2+ hydrate diphosphate
PubChem CID 6096954
SMILES [Mg++].[Mg++].[Mg++].[O-]P([O-])([O-])=O.[O-]P([O-])([O-])=O
View More Specs

Products 2
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10330624
View Documents
Thermo Scientific Acros
391942500
250 g Plastic bottle
£24.22
250g
Please to purchase this item. Need a web account? Register with us today!
10145123
View Documents
Thermo Scientific Acros
391940010
1 kg Plastic bottle
£60.90
1kg
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

53408-95-0
262.86
GVALZJMUIHGIMD-UHFFFAOYSA-H
6096954
Mg3O8P2
MFCD00150251
magnesium phosphate hydrate, trimagnesium diphosphate hydrate, acmc-20akl7, 3mg.2po4.h2o, trimagnesium 2+ hydrate diphosphate
[Mg++].[Mg++].[Mg++].[O-]P([O-])([O-])=O.[O-]P([O-])([O-])=O

Specifications

53408-95-0
Powder
Mg3O8P2
magnesium phosphate hydrate, trimagnesium diphosphate hydrate, acmc-20akl7, 3mg.2po4.h2o, trimagnesium 2+ hydrate diphosphate
GVALZJMUIHGIMD-UHFFFAOYSA-H
trimagnesium(2+) diphosphate
6096954
≥98%
32.0% max. (425°C)
White
98+%
MFCD00150251
Solubility in water: insoluble.
[Mg++].[Mg++].[Mg++].[O-]P([O-])([O-])=O.[O-]P([O-])([O-])=O
262.86
262.86
Pure
Magnesium phosphate, tribasic hydrate
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • Magnesium phosphate, tribasic hydrate

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.