Learn More
Magnesium, 99+%, ribbon 3.2-4.0 mm x 0.3-0.4 mm, Thermo Scientific Chemicals
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Specifications
Color | White |
Melting Point | 651.0°C |
Boiling Point | 1107.0°C |
Density | 1.74 |
Assay Percent Range | 99+% |
Linear Formula | Mg |
UN Number | 1869 |
Quantity | 25 g |
Fieser | 01,627; 02,254; 03,189; 04,315; 05,419; 06,351; 07,218; 10,251; 12,290; 13,170; 15,120; 16,198 |
Flash Point | 500°C |
Show More |
Product Suggestions
Customers who viewed this item also viewed
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
A0466704 | Certificate of Analysis | 10/10/2024 | 413380250,413381000 |
B0149163 | Certificate of Analysis | 14/11/2023 | 413380250,413381000 |
B0149590 | Certificate of Analysis | 14/11/2023 | 413380250,413381000 |
A0457714 | Certificate of Analysis | 10/11/2023 | 413380250,413381000 |
B0147737 | Certificate of Analysis | 20/10/2023 | 413380250,413381000 |
Safety and Handling

- Magnesium
- Danger
- Flammable solid Category 1
- Substance which\, in contact with water\, emits flammable gases Category 2
- Self-heating substance Category 2
- H228-Flammable solid.
- H252-Self-heating in large quantities; may catch fire.
- H261-In contact with water releases flammable gases.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P223-Do not allow contact with water.
Special Offers
-
Multiple Manufacturers
Chemical Extravaganza
Shop now and save up to 97% on top-quality products for your lab needs.Promo Code: 25908Expires: 2025-03-31
-
Multiple Manufacturers
Chromatography and Chemicals Promotion
Explore our new promotion and get up to 70% off some of our most popular products.Promo Code: 24127Expires: 2025-05-31
New Offer Save Up to 70%
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.