missing translation for 'onlineSavingsMsg'
Learn More
Magnesium rod, 6.35mm (0.25in) dia x 25.4mm (1.0in) length, 99.95% (metals basis), Thermo Scientific Chemicals Product Code.: 15455065

Magnesium rod, 6.35mm (0.25in) dia x 25.4mm (1.0in) length, 99.95% (metals basis), Thermo Scientific Chemicals

Product Code. 15455065
25 Pc.
Click to view available options
Quantity:
1 Pc.
25 Pc.
5 Pc.
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

Product Code. 15455065

Brand: Thermo Scientific Alfa Aesar 047073.KM

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7439-95-4
Molecular Formula Mg
Molecular Weight (g/mol) 24.30
MDL Number MFCD00085308
InChI Key JLVVSXFLKOJNIY-UHFFFAOYSA-N
Synonym magnesio, sheet, powdered, magnesio italian, turnings, rieke's active, ribbon, unii-i38zp9992a, hsdb 654, nitrate, acs
PubChem CID 5462224
ChEBI CHEBI:25107
IUPAC Name magnesium
SMILES [Mg]

Specifications

Quantity 25 Pc.
Percent Purity 99.95%
Odor Odorless
Weight ≈1.4g/piece
Assay (metals basis)
Chemical Name or Material Magnesium rod, 6.35mm (0.25 in.) dia. x 25.4mm (1.0 in.) length

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberS13I046 Certificate TypeCertificate of Analysis Date19/08/2022 Catalog Number047073.KF,047073.KJ,047073.KM
Lot NumberM15H005 Certificate TypeCertificate of Analysis Date11/07/2022 Catalog Number047073.KF,047073.KJ,047073.KM
Lot NumberW10H003 Certificate TypeCertificate of Analysis Date29/11/2021 Catalog Number047073.KF,047073.KJ,047073.KM
Lot NumberQ25E011 Certificate TypeCertificate of Analysis Date29/11/2021 Catalog Number047073.KF,047073.KJ,047073.KM
Lot NumberW05A011 Certificate TypeCertificate of Analysis Date24/11/2021 Catalog Number047073.KF,047073.KJ,047073.KM

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Magnesium rod, 6.35mm (0.25in) dia x 25.4mm (1.0in) length, 99.95% (metals basis), Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.